Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTGGAAGACGAGGCCACAGACA[C/G]TTCTGATAGTGAAGGTAATGTGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611193 | ||||||||||||||||||||
Literature Links: |
RRP15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RRP15 - ribosomal RNA processing 15 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016052.3 | 155 | Missense Mutation | ACT,AGT | T,S 42 | NP_057136.2 | |
XM_011509597.2 | 155 | Missense Mutation | ACT,AGT | T,S 42 | XP_011507899.1 |