Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGATCTTAGGAAAGGGCAAGTC[C/T]GGAGAGTGAGTTACCTAATAAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR2M3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR2M3 - olfactory receptor family 2 subfamily M member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004689.1 | 930 | Silent Mutation | TCC,TCT | S,S 310 | NP_001004689.1 |