Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACCACTTACCTATAGGTATCCTG[C/T]GTGGTTTTCTGGTTTAACAGTATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611298 | ||||||||||||||||||||
Literature Links: |
C1orf194 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf194 - chromosome 1 open reading frame 194 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122961.1 | 790 | Silent Mutation | ACA,ACG | T,T 88 | NP_001116433.1 | |
XM_005270448.3 | 790 | Silent Mutation | ACA,ACG | T,T 49 | XP_005270505.1 | |
XM_006710342.3 | 790 | Silent Mutation | ACA,ACG | T,T 199 | XP_006710405.1 | |
XM_006710343.3 | 790 | Silent Mutation | ACA,ACG | T,T 160 | XP_006710406.1 | |
XM_011540647.2 | 790 | Silent Mutation | ACA,ACG | T,T 209 | XP_011538949.1 | |
XM_011540648.1 | 790 | Intron | XP_011538950.1 |
KIAA1324 - KIAA1324 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCARNA2 - small Cajal body-specific RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |