Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCCCTGTCCTCAGGCTTGGCTC[A/G]GCTCAGCAGCTGGGCCTCATGCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612037 | ||||||||||||||||||||
Literature Links: |
MUL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MUL1 - mitochondrial E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024544.2 | 1705 | Nonsense Mutation | CGA,TGA | R,* 289 | NP_078820.2 | |
XM_011542137.2 | 1705 | Nonsense Mutation | CGA,TGA | R,* 250 | XP_011540439.1 |