Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGCCCAGCTGCACCGACTGAAG[A/G]TGGACCAAGCCTGTCACCTGCACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 153390 | ||||||||||||||||||||
Literature Links: |
FAM167B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM167B - family with sequence similarity 167 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032648.2 | 542 | Missense Mutation | ATG,GTG | M,V 113 | NP_116037.2 |
LCK - LCK proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTMR9LP - myotubularin related protein 9-like, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |