Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACTGAGGCCCCATACTCCACGC[A/G]TTATTCTACCCAAGTGGTGAGTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616446 | ||||||||||||||||||||
Literature Links: |
LEXM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LEXM - lymphocyte expansion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110533.1 | 230 | Missense Mutation | CAT,CGT | H,R 59 | NP_001104003.1 | |
NM_152607.2 | 230 | Missense Mutation | CAT,CGT | H,R 59 | NP_689820.2 | |
XM_017000480.1 | 230 | Missense Mutation | CAT,CGT | H,R 59 | XP_016855969.1 | |
XM_017000481.1 | 230 | Missense Mutation | CAT,CGT | H,R 59 | XP_016855970.1 |
TTC22 - tetratricopeptide repeat domain 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |