Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGAAGATATTGTCATTGTTATA[C/G]ATCCTAGTGTGCCAGAAGATGAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616857 | ||||||||||||||||||||
Literature Links: |
CLCA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLCA4 - chloride channel accessory 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012128.3 | 159 | Missense Mutation | CAT,GAT | H,D 39 | NP_036260.2 | |
XM_011541015.2 | 159 | Intron | XP_011539317.1 |