Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCATTTGTCAACCAATCCTGGGCA[C/T]GAGGTCAGGGCTCAGGATGGCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188360 MIM: 188340 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD1B - CD1b molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001764.2 | 419 | Intron | NP_001755.1 | |||
XM_011510119.2 | 419 | Intron | XP_011508421.1 | |||
XM_017002784.1 | 419 | Intron | XP_016858273.1 | |||
XM_017002785.1 | 419 | Intron | XP_016858274.1 | |||
XM_017002786.1 | 419 | Intron | XP_016858275.1 |
CD1C - CD1c molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001765.2 | 419 | Nonsense Mutation | CGA,TGA | R,* 43 | NP_001756.2 | |
XM_005245579.4 | 419 | Nonsense Mutation | CGA,TGA | R,* 43 | XP_005245636.1 |