Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGCTGGTGCAGGGCAGCTTGAC[A/G]GACCTGGTGGAGAGGGCAGAATAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191030 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf189 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf189 - chromosome 1 open reading frame 189 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010979.2 | 266 | Missense Mutation | CGT,TGT | R,C 70 | NP_001010979.1 |
C1orf43 - chromosome 1 open reading frame 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR190B - microRNA 190b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPM3 - tropomyosin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |