Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTAGGGGGATTCCGGGCTAGGT[G/T]CTCTTCCCACTCGACTTCAACCAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611930 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISG20L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ISG20L2 - interferon stimulated exonuclease gene 20 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303095.1 | 1204 | Missense Mutation | AAC,CAC | N,H 345 | NP_001290024.1 | |
XM_005245516.4 | 1204 | Missense Mutation | AAC,CAC | N,H 345 | XP_005245573.1 | |
XM_005245518.4 | 1204 | Missense Mutation | AAC,CAC | N,H 130 | XP_005245575.1 | |
XM_006711560.3 | 1204 | Missense Mutation | AAC,CAC | N,H 345 | XP_006711623.1 | |
XM_006711561.3 | 1204 | Missense Mutation | AAC,CAC | N,H 345 | XP_006711624.1 |
RRNAD1 - ribosomal RNA adenine dimethylase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |