Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGGGTCGCAGCATTGGTCCACAG[A/C]CAGCCAGAGAACCAGGGAATCTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRSS38 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRSS38 - protease, serine 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183062.2 | 108 | Silent Mutation | AGA,AGC | R,S 36 | NP_898885.1 | |
XM_011544175.2 | 108 | Missense Mutation | AGA,AGC | R,S 36 | XP_011542477.1 | |
XM_011544176.2 | 108 | Missense Mutation | AGA,AGC | R,S 36 | XP_011542478.1 |