Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCCGGGCGGGGCCGAGAATTAC[A/G]TAAGACACTGGGCGAGATCTTGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611680 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf116 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf116 - chromosome 1 open reading frame 116 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083924.1 | 1499 | Missense Mutation | CGT,TGT | R,C 278 | NP_001077393.1 | |
NM_023938.5 | 1499 | Missense Mutation | CGT,TGT | R,C 524 | NP_076427.2 | |
XM_005273259.1 | 1499 | Missense Mutation | CGT,TGT | R,C 278 | XP_005273316.1 | |
XM_006711530.1 | 1499 | Missense Mutation | CGT,TGT | R,C 524 | XP_006711593.1 | |
XM_011509973.2 | 1499 | Missense Mutation | CGT,TGT | R,C 278 | XP_011508275.1 |