Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCAGTGAGGATTTCCAGAAAGC[C/T]GAGGATGCCAGCAAGTTTGAGCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613535 MIM: 608458 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0319L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0319L - KIAA0319 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCDN - neurochondrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014839.1 | 755 | Silent Mutation | GCC,GCT | A,A 232 | NP_001014839.1 | |
NM_001014841.1 | 755 | Silent Mutation | GCC,GCT | A,A 215 | NP_001014841.1 | |
NM_014284.2 | 755 | Silent Mutation | GCC,GCT | A,A 232 | NP_055099.1 |