Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCCAGTCCCCCAGGGGCTTATG[C/T]GTCTCCTTGATGACCTGCGGCGACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606811 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH4A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH4A1 - aldehyde dehydrogenase 4 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161504.1 | 1926 | Silent Mutation | ACA,ACG | T,T 490 | NP_001154976.1 | |
NM_001319218.1 | 1926 | Silent Mutation | ACA,ACG | T,T 499 | NP_001306147.1 | |
NM_003748.3 | 1926 | Silent Mutation | ACA,ACG | T,T 550 | NP_003739.2 | |
NM_170726.2 | 1926 | Silent Mutation | ACA,ACG | T,T 550 | NP_733844.1 |