Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCCCCCAAAAACTCCATCTGGG[A/G]CTCTTCATAGAAAAAGGAAAATGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 120830 | ||||||||||||||||||||
Literature Links: |
C4BPA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C4BPA - complement component 4 binding protein alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000715.3 | 354 | Missense Mutation | ACT,GCT | T,A 10 | NP_000706.1 | |
XM_005273251.1 | 354 | Missense Mutation | ACT,GCT | T,A 10 | XP_005273308.1 | |
XM_005273252.4 | 354 | Missense Mutation | ACT,GCT | T,A 10 | XP_005273309.1 |