Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCAGATATAATGTAGCTGAGAT[A/T]GTGATTCATATTCGCAATAAGATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611772 MIM: 603276 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUF2 - NUF2, NDC80 kinetochore complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031423.3 | 342 | Silent Mutation | ATA,ATT | I,I 14 | NP_113611.2 | |
NM_145697.2 | 342 | Silent Mutation | ATA,ATT | I,I 14 | NP_663735.2 | |
XM_011510036.1 | 342 | UTR 5 | XP_011508338.1 |
RGS5 - regulator of G-protein signaling 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |