Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCAGCCAATTCACGATCAACCA[C/T]TACTCTTACTCTCGAAGGGTAGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602105 | ||||||||||||||||||||
Literature Links: |
ASB17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASB17 - ankyrin repeat and SOCS box containing 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080868.2 | 723 | Missense Mutation | ATG,GTG | M,V 195 | NP_543144.1 | |
XM_006710345.3 | 723 | Missense Mutation | ATG,GTG | M,V 147 | XP_006710408.1 | |
XM_006710346.3 | 723 | Missense Mutation | ATG,GTG | M,V 117 | XP_006710409.1 |
MSH4 - mutS homolog 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |