Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCACCAGTAAGGAATATTCAACT[T/C]GAAGGATGTTGCAGCCCAGGATAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606599 | ||||||||||||||||||||
Literature Links: |
TXNIP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TXNIP - thioredoxin interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313972.1 | 1126 | Missense Mutation | CAA,CGA | Q,R 216 | NP_001300901.1 | |
NM_006472.5 | 1126 | Missense Mutation | CAA,CGA | Q,R 271 | NP_006463.3 | |
XM_017000085.1 | 1126 | Missense Mutation | CAA,CGA | Q,R 271 | XP_016855574.1 |