Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATGCTCAGTAGGTGAATCGCAT[C/G]TTTTGTGAGTAGCCCAGTTTGTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607945 MIM: 608341 | ||||||||||||||||||||
Literature Links: |
ADIPOR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADIPOR1 - adiponectin receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CYB5R1 - cytochrome b5 reductase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016243.2 | 971 | Missense Mutation | AAC,AAG | N,K 300 | NP_057327.2 |