Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGCAAAGTCGCCGGCCTTAGTG[A/G]CTGCCACTTCTGACCGCAGCAGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602211 | ||||||||||||||||||||
Literature Links: |
FOXD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FOXD2 - forkhead box D2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004474.3 | 3594 | Missense Mutation | GAC,GGC | D,G 492 | NP_004465.3 |
FOXD2-AS1 - FOXD2 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LINC01389 - long intergenic non-protein coding RNA 1389 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |