Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCCCACCAGATGCCTGTCGAGG[A/G]GACCTCCGTGATGTGGCCAGATCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608507 MIM: 608772 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFN2 - mitofusin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIIP - migration and invasion inhibitory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021933.3 | 363 | Silent Mutation | GGA,GGG | G,G 82 | NP_068752.2 | |
XM_005263487.3 | 363 | Silent Mutation | GGA,GGG | G,G 82 | XP_005263544.1 | |
XM_011541895.1 | 363 | Silent Mutation | GGA,GGG | G,G 82 | XP_011540197.1 | |
XM_011541896.1 | 363 | Silent Mutation | GGA,GGG | G,G 82 | XP_011540198.1 |
MIR6729 - microRNA 6729 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |