Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCTCGCACCAGGAGCTTGGAGA[C/T]GAGGTCTTTGGCTTCACTGGAGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611455 MIM: 606724 | ||||||||||||||||||||
Literature Links: |
KNCN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KNCN - kinocilin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MKNK1 - MAP kinase interacting serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135553.2 | 1186 | Missense Mutation | ATC,GTC | I,V 310 | NP_001129025.1 | |
NM_003684.5 | 1186 | Missense Mutation | ATC,GTC | I,V 351 | NP_003675.2 | |
NM_198973.3 | 1186 | Missense Mutation | ATC,GTC | I,V 310 | NP_945324.1 | |
XM_006711000.1 | 1186 | Missense Mutation | ATC,GTC | I,V 310 | XP_006711063.1 | |
XM_006711001.2 | 1186 | Missense Mutation | ATC,GTC | I,V 304 | XP_006711064.1 | |
XM_006711002.2 | 1186 | Missense Mutation | ATC,GTC | I,V 304 | XP_006711065.1 | |
XM_006711003.1 | 1186 | Missense Mutation | ATC,GTC | I,V 304 | XP_006711066.1 | |
XM_006711006.3 | 1186 | Missense Mutation | ATC,GTC | I,V 255 | XP_006711069.1 | |
XM_006711007.1 | 1186 | Missense Mutation | ATC,GTC | I,V 255 | XP_006711070.1 | |
XM_006711009.3 | 1186 | Missense Mutation | ATC,GTC | I,V 255 | XP_006711072.1 | |
XM_006711013.1 | 1186 | Missense Mutation | ATC,GTC | I,V 215 | XP_006711076.1 | |
XM_006711014.1 | 1186 | Missense Mutation | ATC,GTC | I,V 215 | XP_006711077.1 | |
XM_006711015.1 | 1186 | Missense Mutation | ATC,GTC | I,V 215 | XP_006711078.1 | |
XM_017002652.1 | 1186 | Missense Mutation | ATC,GTC | I,V 313 | XP_016858141.1 | |
XM_017002653.1 | 1186 | Missense Mutation | ATC,GTC | I,V 313 | XP_016858142.1 | |
XM_017002654.1 | 1186 | Missense Mutation | ATC,GTC | I,V 313 | XP_016858143.1 | |
XM_017002655.1 | 1186 | Missense Mutation | ATC,GTC | I,V 298 | XP_016858144.1 | |
XM_017002656.1 | 1186 | Missense Mutation | ATC,GTC | I,V 298 | XP_016858145.1 | |
XM_017002657.1 | 1186 | Missense Mutation | ATC,GTC | I,V 255 | XP_016858146.1 | |
XM_017002658.1 | 1186 | Missense Mutation | ATC,GTC | I,V 313 | XP_016858147.1 | |
XM_017002659.1 | 1186 | Missense Mutation | ATC,GTC | I,V 310 | XP_016858148.1 | |
XM_017002660.1 | 1186 | Missense Mutation | ATC,GTC | I,V 304 | XP_016858149.1 | |
XM_017002661.1 | 1186 | Missense Mutation | ATC,GTC | I,V 304 | XP_016858150.1 | |
XM_017002662.1 | 1186 | Missense Mutation | ATC,GTC | I,V 304 | XP_016858151.1 | |
XM_017002663.1 | 1186 | Missense Mutation | ATC,GTC | I,V 298 | XP_016858152.1 | |
XM_017002664.1 | 1186 | Missense Mutation | ATC,GTC | I,V 215 | XP_016858153.1 | |
XM_017002665.1 | 1186 | Missense Mutation | ATC,GTC | I,V 255 | XP_016858154.1 | |
XM_017002666.1 | 1186 | Missense Mutation | ATC,GTC | I,V 215 | XP_016858155.1 | |
XM_017002667.1 | 1186 | Missense Mutation | ATC,GTC | I,V 215 | XP_016858156.1 | |
XM_017002668.1 | 1186 | Intron | XP_016858157.1 |
MKNK1-AS1 - MKNK1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |