Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCAAGTTTCCAAGAGATATGAA[C/T]GGTATCAGAGGTGACAGACTTCACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604808 MIM: 611567 | ||||||||||||||||||||
Literature Links: |
FLRT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLRT3 - fibronectin leucine rich transmembrane protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013281.3 | 2432 | Missense Mutation | ATT,GTT | I,V 423 | NP_037413.1 | |
NM_198391.2 | 2432 | Missense Mutation | ATT,GTT | I,V 423 | NP_938205.1 | |
XM_005260682.4 | 2432 | Missense Mutation | ATT,GTT | I,V 423 | XP_005260739.1 | |
XM_011529204.2 | 2432 | Missense Mutation | ATT,GTT | I,V 423 | XP_011527506.1 | |
XM_011529205.2 | 2432 | Missense Mutation | ATT,GTT | I,V 423 | XP_011527507.1 |
MACROD2 - MACRO domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033087.1 | 2432 | Intron | NP_001028259.1 | |||
NM_080676.5 | 2432 | Intron | NP_542407.2 | |||
XM_017027675.1 | 2432 | Intron | XP_016883164.1 | |||
XM_017027676.1 | 2432 | Intron | XP_016883165.1 | |||
XM_017027677.1 | 2432 | Intron | XP_016883166.1 |