Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCGGTGTCCTCTTCTTCCACAG[C/T]GCTCCTGTTGACACAAACAGCCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601415 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTSF1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GTSF1L - gametocyte specific factor 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008901.1 | 510 | Missense Mutation | ACT,GCT | T,A 68 | NP_001008901.1 | |
NM_176791.3 | 510 | Missense Mutation | ACT,GCT | T,A 68 | NP_789761.1 | |
XM_005260298.4 | 510 | Missense Mutation | ACT,GCT | T,A 68 | XP_005260355.1 |
LOC101927200 - uncharacterized LOC101927200 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYBL2 - MYB proto-oncogene like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |