Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTGGGTTTCTCGTCGCAGGTGT[G/T]CAGGTACTCCCTGCAGAAGCTGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613110 MIM: 603106 | ||||||||||||||||||||
Literature Links: |
BLCAP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BLCAP - bladder cancer associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167820.1 | 203 | Intron | NP_001161292.1 | |||
NM_001167821.1 | 203 | Intron | NP_001161293.1 | |||
NM_001167822.2 | 203 | Intron | NP_001161294.1 | |||
NM_001167823.1 | 203 | Intron | NP_001161295.1 | |||
NM_001317074.1 | 203 | Intron | NP_001304003.1 | |||
NM_001317075.1 | 203 | Intron | NP_001304004.1 | |||
NM_006698.3 | 203 | Intron | NP_006689.1 | |||
XM_017027599.1 | 203 | Intron | XP_016883088.1 |
NNAT - neuronatin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322802.1 | 203 | Silent Mutation | GTG,GTT | V,V 51 | NP_001309731.1 | |
NM_005386.3 | 203 | Missense Mutation | TGC,TTC | C,F 53 | NP_005377.1 | |
NM_181689.2 | 203 | Missense Mutation | TGC,TTC | C,F 26 | NP_859017.1 |