Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGAGTTCTTACCCCATCATCAGT[C/G]TGGACCTTTCTTCTCTTCTTGACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610952 MIM: 610833 | ||||||||||||||||||||
Literature Links: |
CRNKL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRNKL1 - crooked neck pre-mRNA splicing factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278625.1 | 2399 | Missense Mutation | CAC,CAG | H,Q 777 | NP_001265554.1 | |
NM_001278626.1 | 2399 | Missense Mutation | CAC,CAG | H,Q 505 | NP_001265555.1 | |
NM_001278627.1 | 2399 | Missense Mutation | CAC,CAG | H,Q 505 | NP_001265556.1 | |
NM_001278628.1 | 2399 | Missense Mutation | CAC,CAG | H,Q 628 | NP_001265557.1 | |
NM_016652.5 | 2399 | Missense Mutation | CAC,CAG | H,Q 789 | NP_057736.4 |
NAA20 - N(alpha)-acetyltransferase 20, NatB catalytic subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |