Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCTTTTAAAATTGTATTTCCAG[C/G]TTAAAAGATGAAATCACATCTGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
IFT52 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFT52 - intraflagellar transport 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303458.2 | 543 | Missense Mutation | AGC,AGG | S,R 40 | NP_001290387.1 | |
NM_001303459.2 | 543 | Missense Mutation | AGC,AGG | S,R 40 | NP_001290388.1 | |
NM_001323578.1 | 543 | UTR 5 | NP_001310507.1 | |||
NM_001323579.1 | 543 | UTR 5 | NP_001310508.1 | |||
NM_001323580.1 | 543 | UTR 5 | NP_001310509.1 | |||
NM_001323581.1 | 543 | UTR 5 | NP_001310510.1 | |||
NM_016004.4 | 543 | Missense Mutation | AGC,AGG | S,R 40 | NP_057088.2 | |
XM_017027863.1 | 543 | UTR 5 | XP_016883352.1 |