Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATTAGGTCGACAAAGATGCTG[A/G]ATTAGTGGCCCAATGGAACTATTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RTFDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RTFDC1 - replication termination factor 2 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283035.1 | 199 | Missense Mutation | GAA,GGA | E,G 59 | NP_001269964.1 | |
NM_001283036.1 | 199 | Missense Mutation | GAA,GGA | E,G 29 | NP_001269965.1 | |
NM_001283037.1 | 199 | Missense Mutation | GAA,GGA | E,G 29 | NP_001269966.1 | |
NM_016407.4 | 199 | Missense Mutation | GAA,GGA | E,G 29 | NP_057491.2 | |
XM_017027872.1 | 199 | Missense Mutation | GAA,GGA | E,G 10 | XP_016883361.1 |