Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGACATGGGCAGGTTCGAGGCA[C/T]GCACCACAGCCTCACGTGTGGCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604205 MIM: 603038 | ||||||||||||||||||||
Literature Links: |
CPNE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPNE1 - copine 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198863.1 | 1507 | Missense Mutation | CAT,CGT | H,R 436 | NP_001185792.1 | |
NM_003915.5 | 1507 | Missense Mutation | CAT,CGT | H,R 442 | NP_003906.2 | |
NM_152925.2 | 1507 | Missense Mutation | CAT,CGT | H,R 437 | NP_690902.1 | |
NM_152926.2 | 1507 | Missense Mutation | CAT,CGT | H,R 437 | NP_690903.1 | |
NM_152927.2 | 1507 | Missense Mutation | CAT,CGT | H,R 437 | NP_690904.1 | |
NM_152928.2 | 1507 | Missense Mutation | CAT,CGT | H,R 437 | NP_690905.1 |
SPAG4 - sperm associated antigen 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |