Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGGAGAAGGCCTCCACGTAGA[C/T]GAGGTAGTTGTGGGGACAGTGCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613979 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRPF6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRPF6 - pre-mRNA processing factor 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAMD10 - sterile alpha motif domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080621.4 | 584 | Missense Mutation | ATC,GTC | I,V 137 | NP_542188.1 | |
XM_005260199.4 | 584 | Missense Mutation | ATC,GTC | I,V 176 | XP_005260256.1 | |
XM_006723705.3 | 584 | Missense Mutation | ATC,GTC | I,V 143 | XP_006723768.1 | |
XM_011528565.2 | 584 | Missense Mutation | ATC,GTC | I,V 341 | XP_011526867.1 | |
XM_017027671.1 | 584 | Missense Mutation | ATC,GTC | I,V 137 | XP_016883160.1 |
ZNF512B - zinc finger protein 512B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |