Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCGGGAGATGGCGAGGCGGGCC[C/T]CCAACAGGCGGGTAGGAGCCCCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608802 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
L3MBTL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
L3MBTL1 - l(3)mbt-like 1 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015478.6 | 330 | Missense Mutation | CCC,CTC | P,L 74 | NP_056293.4 | |
NM_032107.4 | 330 | Missense Mutation | CCC,CTC | P,L 142 | NP_115479.4 |