Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGGTGTAGGTAAATCGAGTATT[C/G]TGTGGCGGTTTGTGGAAGACAGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612966 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP4R1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP4R1L - protein phosphatase 4 regulatory subunit 1 like (pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB22A - RAB22A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020673.2 | 326 | Missense Mutation | CTG,GTG | L,V 22 | NP_065724.1 |