Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCGGTTCTTCTCAATCACCTCT[C/T]GCAGCCCTTTGGCAAAGTGGAGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613111 MIM: 608597 MIM: 172425 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTSA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTSA - cathepsin A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000308.3 | 1418 | Intron | NP_000299.2 | |||
NM_001127695.2 | 1418 | Intron | NP_001121167.1 | |||
NM_001167594.2 | 1418 | Intron | NP_001161066.1 |
NEURL2 - neuralized E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLTP - phospholipid transfer protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242920.1 | 1418 | Missense Mutation | CAA,CGA | Q,R 374 | NP_001229849.1 | |
NM_001242921.1 | 1418 | Missense Mutation | CAA,CGA | Q,R 381 | NP_001229850.1 | |
NM_006227.3 | 1418 | Missense Mutation | CAA,CGA | Q,R 469 | NP_006218.1 | |
NM_182676.2 | 1418 | Missense Mutation | CAA,CGA | Q,R 417 | NP_872617.1 |