Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCGAAAACAGACAGGTGATCCTC[C/T]TGCAGGTGCCCGTGCGCCCGGCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600010 | ||||||||||||||||||||
Literature Links: |
CFAP61 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CFAP61 - cilia and flagella associated protein 61 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
INSM1 - INSM transcriptional repressor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002196.2 | 1651 | Silent Mutation | CTG,TTG | L,L 502 | NP_002187.1 |