Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGCAGCCAAGTTTTGCTACTTAC[A/G]GGAGTACTGAAGGCCCTCGGTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604612 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NKX2-2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NKX2-2 - NK2 homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002509.3 | 616 | Silent Mutation | CTG,TTG | L,L 87 | NP_002500.1 | |
XM_006723566.3 | 616 | Intron | XP_006723629.1 |