Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCGGAATGGAGACTTCGAGATGT[A/G]CAGGCCCTTCGTGACATGGTGCGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TTPAL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TTPAL - alpha tocopherol transfer protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039199.2 | 310 | Silent Mutation | GTA,GTG | V,V 58 | NP_001034288.1 | |
NM_001261839.1 | 310 | Silent Mutation | GTA,GTG | V,V 58 | NP_001248768.1 | |
NM_024331.4 | 310 | Silent Mutation | GTA,GTG | V,V 58 | NP_077307.2 | |
XM_011529043.2 | 310 | Silent Mutation | GTA,GTG | V,V 58 | XP_011527345.1 | |
XM_011529044.2 | 310 | Silent Mutation | GTA,GTG | V,V 58 | XP_011527346.1 | |
XM_011529045.2 | 310 | Silent Mutation | GTA,GTG | V,V 58 | XP_011527347.1 | |
XM_017028063.1 | 310 | Silent Mutation | GTA,GTG | V,V 58 | XP_016883552.1 |