Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGCGCCGCCATGGCTCTGCTGTC[C/T]GAGGGCCTGGACGAGGTGCCCGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613275 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM83D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM83D - family with sequence similarity 83 member D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030919.2 | 146 | Silent Mutation | TCC,TCT | S,S 5 | NP_112181.3 | |
XM_017028088.1 | 146 | Intron | XP_016883577.1 |
PPP1R16B - protein phosphatase 1 regulatory subunit 16B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |