Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCTCCCCAGGCAGGTAGAGTCA[A/G]TGTGTCGGCTGCAGCAGCAGGCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614824 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP5S1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP5S1 - adaptor related protein complex 5 sigma 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204446.1 | 441 | Missense Mutation | ATG,GTG | M,V 64 | NP_001191375.1 | |
NM_001204447.1 | 441 | Missense Mutation | ATG,GTG | M,V 64 | NP_001191376.1 | |
NM_018347.2 | 441 | Missense Mutation | ATG,GTG | M,V 64 | NP_060817.1 |