Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTGAAGGTGCCGAAGATGATCA[A/G]GCAGAAGACGAAGCACTCCCAGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610994 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM189 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM189 - transmembrane protein 189 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162505.1 | 684 | Silent Mutation | CTG,TTG | L,L 172 | NP_001155977.1 | |
NM_199129.2 | 684 | Silent Mutation | CTG,TTG | L,L 175 | NP_954580.1 |
TMEM189-UBE2V1 - TMEM189-UBE2V1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199203.2 | 684 | Silent Mutation | CTG,TTG | L,L 175 | NP_954673.1 |