Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCAGAGCAGCTTGGTGAAGGCC[C/T]GGCGGAAGCTGTGGTGGCACAGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604525 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HRH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HRH3 - histamine receptor H3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007232.2 | 1262 | Missense Mutation | NP_009163.2 | |||
XM_005260266.3 | 1262 | Missense Mutation | XP_005260323.1 | |||
XM_017027623.1 | 1262 | Missense Mutation | XP_016883112.1 |