Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCGTCTTCTGCTCCTCAGTCAGG[C/G]CGGCCCCCTCCACAGTGAAGGTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190196 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TGM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TGM2 - transglutaminase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323316.1 | 2097 | Missense Mutation | GCC,GGC | A,G 628 | NP_001310245.1 | |
NM_001323317.1 | 2097 | Missense Mutation | GCC,GGC | A,G 547 | NP_001310246.1 | |
NM_001323318.1 | 2097 | Missense Mutation | GCC,GGC | A,G 568 | NP_001310247.1 | |
NM_004613.3 | 2097 | Missense Mutation | GCC,GGC | A,G 628 | NP_004604.2 | |
NM_198951.2 | 2097 | Intron | NP_945189.1 | |||
XM_011529028.1 | 2097 | Missense Mutation | GCC,GGC | A,G 628 | XP_011527330.1 |