Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGTGCTTCTTCTCCTTGGCACC[A/G]CCTGCTACCCGGCGTGAGTGGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604866 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLAGL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLAGL2 - PLAG1 like zinc finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002657.3 | 1614 | Intron | NP_002648.1 | |||
XM_005260436.3 | 1614 | Silent Mutation | GGC,GGT | G,G 184 | XP_005260493.1 | |
XM_011528863.2 | 1614 | Silent Mutation | GGC,GGT | G,G 184 | XP_011527165.1 | |
XM_011528864.2 | 1614 | Silent Mutation | GGC,GGT | G,G 184 | XP_011527166.1 |
TSPY26P - testis specific protein, Y-linked 26, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |