Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGTATACCTCATTTCTCGAAGAC[C/G]CTTCACATGCTGAATGCATTTCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604205 MIM: 603485 MIM: 607179 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPNE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPNE1 - copine 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFS1 - NFS1 cysteine desulfurase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198989.1 | 1218 | Missense Mutation | CGT,GGT | R,G 381 | NP_001185918.1 | |
NM_021100.4 | 1218 | Missense Mutation | CGT,GGT | R,G 432 | NP_066923.3 |
RBM12 - RNA binding motif protein 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |