Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTCCAGGCATCCACTTCCTCCA[C/T]TACAGCTCTGTGGACAAGGCTAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602879 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AAR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AAR2 - AAR2 splicing factor homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271874.1 | 402 | Silent Mutation | CAC,CAT | H,H 59 | NP_001258803.1 | |
NM_015511.4 | 402 | Silent Mutation | CAC,CAT | H,H 59 | NP_056326.2 | |
XM_006723770.3 | 402 | Silent Mutation | CAC,CAT | H,H 59 | XP_006723833.1 | |
XM_011528762.2 | 402 | Silent Mutation | CAC,CAT | H,H 59 | XP_011527064.1 | |
XM_011528763.2 | 402 | Silent Mutation | CAC,CAT | H,H 59 | XP_011527065.1 |
EPB41L1 - erythrocyte membrane protein band 4.1 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |