Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTTACGGCAGAGGGGCCTGGACT[C/T]GGTCCCGCCCGCCTCCAGAGCAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605299 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCOA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCOA6 - nuclear receptor coactivator 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TP53INP2 - tumor protein p53 inducible nuclear protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005260476.2 | 222 | Silent Mutation | CTC,CTT | L,L 86 | XP_005260533.1 | |
XM_006723841.2 | 222 | Silent Mutation | CTC,CTT | L,L 20 | XP_006723904.1 | |
XM_011528952.2 | 222 | Silent Mutation | CTC,CTT | L,L 86 | XP_011527254.1 | |
XM_011528953.2 | 222 | Silent Mutation | CTC,CTT | L,L 86 | XP_011527255.1 |