Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGAATCTGTAAGAGAATTTCTG[G/T]ATTTTGCCCGAGGAGTCCATGGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CASS4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CASS4 - Cas scaffolding protein family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164114.1 | 2108 | Missense Mutation | GAT,TAT | D,Y 441 | NP_001157586.1 | |
NM_001164115.1 | 2108 | Intron | NP_001157587.1 | |||
NM_001164116.1 | 2108 | Missense Mutation | GAT,TAT | D,Y 495 | NP_001157588.1 | |
NM_020356.3 | 2108 | Missense Mutation | GAT,TAT | D,Y 495 | NP_065089.2 | |
XM_006723831.3 | 2108 | Missense Mutation | GAT,TAT | D,Y 441 | XP_006723894.1 |