Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTTAGCACAAACCGGCTAGGGG[C/T]AGCAAGGGCCTCGGCGATGGCCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605832 MIM: 601002 | ||||||||||||||||||||
Literature Links: |
ACSS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACSS2 - acyl-CoA synthetase short-chain family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GSS - glutathione synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000178.3 | 1168 | Missense Mutation | ACC,GCC | T,A 357 | NP_000169.1 | |
NM_001322494.1 | 1168 | Missense Mutation | ACC,GCC | T,A 357 | NP_001309423.1 | |
NM_001322495.1 | 1168 | Missense Mutation | ACC,GCC | T,A 357 | NP_001309424.1 |