Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCCAGCCCCTTCAAGGCCTTGG[A/G]GCCACCATCACCTTGGGATAGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC35C2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC35C2 - solute carrier family 35 member C2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281457.1 | 1239 | Missense Mutation | CCC,TCC | P,S 213 | NP_001268386.1 | |
NM_001281458.1 | 1239 | Missense Mutation | CCC,TCC | P,S 356 | NP_001268387.1 | |
NM_001281459.1 | 1239 | Missense Mutation | CCC,TCC | P,S 192 | NP_001268388.1 | |
NM_001281460.1 | 1239 | Missense Mutation | CCC,TCC | P,S 327 | NP_001268389.1 | |
NM_015945.11 | 1239 | Missense Mutation | CCC,TCC | P,S 327 | NP_057029.8 | |
NM_173073.3 | 1239 | Missense Mutation | CCC,TCC | P,S 306 | NP_775096.1 | |
NM_173179.3 | 1239 | Missense Mutation | CCC,TCC | P,S 327 | NP_775271.1 | |
XM_011528831.1 | 1239 | Missense Mutation | CCC,TCC | P,S 327 | XP_011527133.1 | |
XM_011528832.1 | 1239 | Missense Mutation | CCC,TCC | P,S 327 | XP_011527134.1 | |
XM_011528833.1 | 1239 | Missense Mutation | CCC,TCC | P,S 327 | XP_011527135.1 | |
XM_011528834.1 | 1239 | Missense Mutation | CCC,TCC | P,S 326 | XP_011527136.1 | |
XM_011528835.1 | 1239 | Missense Mutation | CCC,TCC | P,S 289 | XP_011527137.1 | |
XM_011528836.1 | 1239 | Missense Mutation | CCC,TCC | P,S 213 | XP_011527138.1 | |
XM_011528837.1 | 1239 | Missense Mutation | CCC,TCC | P,S 213 | XP_011527139.1 | |
XM_011528838.1 | 1239 | Missense Mutation | CCC,TCC | P,S 175 | XP_011527140.1 | |
XM_017027856.1 | 1239 | Missense Mutation | CCC,TCC | P,S 327 | XP_016883345.1 | |
XM_017027857.1 | 1239 | Missense Mutation | CCC,TCC | P,S 327 | XP_016883346.1 | |
XM_017027858.1 | 1239 | Missense Mutation | CCC,TCC | P,S 327 | XP_016883347.1 | |
XM_017027859.1 | 1239 | Missense Mutation | CCC,TCC | P,S 327 | XP_016883348.1 | |
XM_017027860.1 | 1239 | Missense Mutation | CCC,TCC | P,S 327 | XP_016883349.1 | |
XM_017027861.1 | 1239 | Missense Mutation | CCC,TCC | P,S 306 | XP_016883350.1 | |
XM_017027862.1 | 1239 | UTR 3 | XP_016883351.1 |