Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGGCTTTGAGACTTGCTTGGGG[A/G]ACTTGGGTAGCGTCAGAGGAGTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605267 | ||||||||||||||||||||
Literature Links: |
JPH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
JPH2 - junctophilin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OSER1 - oxidative stress responsive serine rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016470.7 | 812 | Silent Mutation | GTC,GTT | V,V 172 | NP_057554.4 | |
XM_011528851.1 | 812 | Silent Mutation | GTC,GTT | V,V 172 | XP_011527153.1 | |
XM_011528853.2 | 812 | Silent Mutation | GTC,GTT | V,V 172 | XP_011527155.1 | |
XM_011528854.2 | 812 | Silent Mutation | GTC,GTT | V,V 134 | XP_011527156.1 | |
XM_017027873.1 | 812 | Silent Mutation | GTC,GTT | V,V 172 | XP_016883362.1 |