Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCCCCTTGGATGGACTTTTTGA[C/T]GCGAAGCAACATGGTGGTCAGCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613842 MIM: 611270 | ||||||||||||||||||||
Literature Links: |
GZF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GZF1 - GDNF inducible zinc finger protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAPB - NSF attachment protein beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283018.1 | 976 | Missense Mutation | ATC,GTC | I,V 287 | NP_001269947.1 | |
NM_001283020.1 | 976 | Missense Mutation | ATC,GTC | I,V 244 | NP_001269949.1 | |
NM_001283026.1 | 976 | Missense Mutation | ATC,GTC | I,V 189 | NP_001269955.1 | |
NM_022080.2 | 976 | Missense Mutation | ATC,GTC | I,V 283 | NP_071363.1 | |
XM_011529313.1 | 976 | Missense Mutation | ATC,GTC | I,V 198 | XP_011527615.1 | |
XM_011529315.2 | 976 | Intron | XP_011527617.1 | |||
XM_017028008.1 | 976 | Missense Mutation | ATC,GTC | I,V 198 | XP_016883497.1 |